Learn More
ENTPD2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ENTPD2.
Brand: Abnova PAB26062.50ug
Description
The protein encoded by this gene is the type 2 enzyme of the ecto-nucleoside triphosphate diphosphohydrolase family (E-NTPDase). E-NTPDases are a family of ecto-nucleosidases that hydrolyze 5'-triphosphates. This ecto-ATPase is an integral membrane protein. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq]
Specifications
| ENTPD2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ENTPD2. | |
| In PBS (0.09% sodium azide) | |
| Q9Y5L3 | |
| ENTPD2 | |
| A synthetic peptide corresponding to 18 amino acids at internal region of human ENTPD2. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| ENTPD2 | |
| CD39L1/NTPDase-2 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 954 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only