Learn More
EMR3 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of EMR3.
Brand: Abnova PAB16404.50ug
Description
This gene encodes a member of the class B seven-span transmembrane (TM7) receptor family expressed predominantly by cells of the immune system. Family members are characterized by an extended extracellular region with a variable number of N-terminal epidermal growth factor (EGF)-like domains coupled to a TM7 domain via a mucin-like spacer domain. This gene is closely linked to the gene encoding egf-like molecule containing mucin-like hormone receptor 2 on chromosome 19. This protein may play a role in myeloid-myeloid interactions during immune and inflammatory responses. [provided by RefSeq
Specifications
| EMR3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of EMR3. | |
| In PBS (0.09% sodium azide) | |
| Q9BY15 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 84658 | |
| Human, Monkey | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| EMR3 | |
| EMR3 | |
| A synthetic peptide (conjugated with KLH) corresponding to human EMR3. | |
| 50 μg | |
| Primary | |
| C-terminal Cytoplasmic domain of human. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only