Learn More
EMR2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of EMR2.
Brand: Abnova PAB24986.100uL
Description
This gene encodes a member of the class B seven-span transmembrane (TM7) receptor family expressed predominantly by cells of the immune system. Family members are characterized by an extended extracellular region with a variable number of N-terminal epidermal growth factor (EGF)-like domains coupled to a TM7 domain via a mucin-like spacer domain. This gene is closely linked to the gene encoding egf-like molecule containing mucin-like hormone receptor 3 on chromosome 19. This protein does not interact with the ligand decay accelerating factor for complement, unlike the related CD97 antigen, and indicates that these very closely related proteins likely have nonredundant functions. This gene results in multiple transcripts encoding distinct isoforms. [provided by RefSeq
Specifications
| EMR2 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| EMR2 | |
| EMR2 | |
| A synthetic peptide corresponding to EMR2. | |
| 100 μL | |
| Primary | |
| EMR2 polyclonal antibody detects endogenous levels of EMR2 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of EMR2. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| CD312/DKFZp781B135 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 30817 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only