missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELOVL3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ELOVL3.
Brand: Abnova PAB17713.100ug
This item is not returnable.
View return policy
Description
ELOVL3 plays a role in fatty acid chain elongation and formation of neutral lipids required for skin barrier function (Westerberg et al., 2004 [PubMed 14581464]).[supplied by OMIM
Specifications
| ELOVL3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ELOVL3. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| Q9HB03 | |
| ELOVL3 | |
| A synthetic peptide corresponding to internal of human ELOVL3. | |
| RUO | |
| 83401 | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
| ELOVL3 | |
| CIG-30/CIG30/MGC21435 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total ELOVL3 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction