missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELL, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ELL.
Brand: Abnova PAB26864.100uL
This item is not returnable.
View return policy
Specifications
| ELL | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| ELL | |
| C19orf17/DKFZp434I1916/ELL1/Men | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 8178 | |
| Human, Mouse | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of ELL. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| P55199 | |
| ELL | |
| A synthetic peptide corresponding to human ELL. | |
| 100 μL | |
| Primary | |
| ELL polyclonal antibody detects endogenous levels of ELL protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction