Learn More
EIF3K Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant EIF3K.
Brand: Abnova PAB24415.100uL
Description
The 700kDa eukaryotic translation initiation factor-3 (eIF3) is the largest eIF and contains at least 12 subunits, including EIF2S12. eIF3 plays an essential role in translation by binding directly to the 40S ribosomal subunit and promoting formation of the 40S preinitiation complex (Mayeur et al., 2003 [PubMed 14519125]).[supplied by OMIM
Sequence: MAMFEQMRANVGKLLKGIDRYNPENLATLERYVETQAKENAYDLEANLAVLKLYQFNPAFFQTTVTAQILLKASpecifications
| EIF3K | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant EIF3K. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9UBQ5 | |
| EIF3K | |
| Recombinant protein corresponding to amino acids of human EIF3K. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Western Blot, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| EIF3K | |
| ARG134/EIF3-p28/EIF3S12/HSPC029/M9/MSTP001/PLAC-24/PLAC24/PRO1474/PTD001 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 27335 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only