missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF2AK3 (phospho T982), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human EIF2AK3.
Brand: Abnova PAB29217.100uL
This item is not returnable.
View return policy
Specifications
| EIF2AK3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human EIF2AK3. | |
| In PBS, pH 7.4 (without mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide) | |
| Q9NZJ5 | |
| EIF2AK3 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T982 of human EIF2AK3. | |
| 100 μL | |
| Primary | |
| This antibody detects endogenous levels of EIF2AK3 only when phosphorylated at threonine 982. | |
| Store at -20°C. Avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| EIF2AK3 | |
| DKFZp781H1925/HRI/PEK/PERK/WRS | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 9451 | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction