Learn More
EHMT1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of EHMT1.
Brand: Abnova PAB2182.400uL
Description
The protein encoded by this gene is a histone methyltransferase that is part of the E2F6 complex, which represses transcription. The encoded protein methylates the Lys-9 position of histone H3, which tags it for transcriptional repression. This protein may be involved in the silencing of MYC- and E2F-responsive genes and therefore could play a role in the G0/G1 cell cycle transition. Defects in this gene are a cause of chromosome 9q subtelomeric deletion syndrome (9q-syndrome). Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| EHMT1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of EHMT1. | |
| In PBS (0.09% sodium azide) | |
| NP_079033;Q9H9B1 | |
| EHMT1 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human EHMT1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| EHMT1 | |
| DEL9q34/DKFZp667M072/EUHMTASE1/Eu-HMTase1/FLJ12879/FP13812/GLP/KIAA1876/KMT1D/bA188C12.1 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 79813 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only