Learn More
EGFR (phospho Y1172), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of EGFR.
Brand: Abnova PAB25345.100ug
Description
The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation. Mutations in this gene are associated with lung cancer. [provided by RefSeq
Sequence: P-D-Yp-Q-QSpecifications
| EGFR | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of EGFR. | |
| In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| P00533 | |
| EGFR | |
| Synthetic phosphopeptide corresponding to residues surrounding Y1172 of human EGFR. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| EGFR | |
| ERBB/ERBB1/HER1/PIG61/mENA | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 1956 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only