Learn More
EGFR (phospho Y1197), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of EGFR.
Brand: Abnova PAB10009.100ug
Description
The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation. Mutations in this gene are associated with lung cancer. [provided by RefSeq
Specifications
| EGFR | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of EGFR. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| P00533;NP_005219 | |
| EGFR | |
| Synthetic phosphopeptide corresponding to residues surrounding Y1197 of human EGFR. | |
| RUO | |
| 1956 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:4000-1:20000) Western Blot (1:250-1:1500) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| EGFR | |
| ERBB/ERBB1/HER1/PIG61/mENA | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Reactivity occurs against human EGFR pY1197 protein and This antibody is specific to the phosphorylated form of the protein. Reactivity with non-phosphorylated human EGFR is minimal by ELISA. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only