missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EFNB1/EFNB2/EFNB3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human EFNB1/EFNB2/EFNB3.
Brand: Abnova PAB29232.100uL
This item is not returnable.
View return policy
Specifications
| EFNB1/EFNB2/EFNB3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human EFNB1/EFNB2/EFNB3. | |
| In PBS, pH 7.4 (without mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide) | |
| CFND/CFNS/EFL3/EPLG2/Elk-L/LERK2/MGC8782 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 1947/1948/1949 | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Western Blot (1:500-1000) The optimal working dilution should be determined by the end user. | |
| P98172;P52799;Q15768 | |
| EFNB1 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y324 of human EFNB1/EFNB2/EFNB3. | |
| 100 μL | |
| Primary | |
| This antibody detects endogenous levels of EFNB1/EFNB2/EFNB3 only when phosphorylated at tyrosine 324. | |
| Store at -20°C. Avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction