Learn More
EDA2R, Rabbit, Polyclonal Antibody, Abnova™
Rabbit Polyclonal antibody raised against recombinant EDA2R.
Brand: Abnova PAB12963.100ug
Description
EDA-A1 and EDA-A2 are two isoforms of ectodysplasin that are encoded by the anhidrotic ectodermal dysplasia (EDA) gene. Mutations in EDA give rise to a clinical syndrome characterized by loss of hair, sweat glands, and teeth. The protein encoded by this gene specifically binds to EDA-A2 isoform. This protein is a type III transmembrane protein of the TNFR (tumor necrosis factor receptor) superfamily, and contains 3 cysteine-rich repeats and a single transmembrane domain but lacks an N-terminal signal peptide. Multiple alternatively spliced transcript variants have been found for this gene, but some variants lack sufficient support. [provided by RefSeq
Specifications
| EDA2R | |
| Polyclonal | |
| Rabbit Polyclonal antibody raised against recombinant EDA2R. | |
| In PBS (0.02% sodium azide) | |
| AAQ89953 | |
| EDA2R | |
| Recombinant protein corresponding to human EDA2R. | |
| RUO | |
| 60401 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| EDA2R | |
| EDA-A2R/EDAA2R/TNFRSF27/XEDAR | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only