missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECE1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ECE1.
Brand: Abnova PAB28442.100uL
This item is not returnable.
View return policy
Description
Endothelin-converting enzyme-1 is involved in the proteolytic processing of endothelin-1 (EDN1; MIM 131240), -2 (EDN2; MIM 131241), and -3 (EDN3; MIM 131242) to biologically active peptides.[supplied by OMIM
Sequence: IMDPKELDKVFNDYTAVPDLYFENAMRFFNFSWRVTADQLRKAPNRDQWSMTPPMVNAYYSPTKNEIVFPAGILQAPFYTRSSPKALNFGGIGVVVGHELTHAFDDQGREYDKDGNLRPWWKNSSVEAFKRSpecifications
| ECE1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ECE1. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| B4DKB2 | |
| ECE1 | |
| Recombinant protein corresponding to amino acids of recombinant ECE1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry(1:20-1:50) The optimal working dilution should be determined by the end user. | |
| ECE1 | |
| ECE | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1889 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur