Learn More
EBI3 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of EBI3.
Brand: Abnova PAB16680.100ug
Description
This gene was identified by its induced expression in B lymphocytes in response Epstein-Barr virus infection. It encodes a secreted glycoprotein belonging to the hematopoietin receptor family, and heterodimerizes with a 28kDa protein to form interleukin 27 (IL-27). IL-27 regulates T cell and inflammatory responses, in part by activating the Jak/STAT pathway of CD4+ T cells. [provided by RefSeq]
Specifications
| EBI3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of EBI3. | |
| In PBS (0.02% sodium azide) | |
| NP_005746 | |
| EBI3 | |
| A synthetic peptide corresponding to C-terminus 16 amino acids from of human EBI3. | |
| RUO | |
| 10148 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| EBI3 | |
| IL27B | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only