missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of E4.
Brand: Abnova PAB16542.100ug
This item is not returnable.
View return policy
Specifications
| E4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of E4. | |
| In PBS (0.02% sodium azide) | |
| P89079 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Adenovirus | |
| Liquid |
| ELISA | |
| Unconjugated | |
| ELISA (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| E4 | |
| E4 | |
| A synthetic peptide corresponding to C-terminus 16 amino acids of Adenovirus E4. | |
| RUO | |
| 6386282 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction