Learn More
E2F1 (phospho S337), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of E2F1.
Brand: Abnova PAB0449.400uL
Description
The protein encoded by this gene is a member of the E2F family of transcription factors. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionally conserved domains found in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. This protein and another 2 members, E2F2 and E2F3, have an additional cyclin binding domain. This protein binds preferentially to retinoblastoma protein pRB in a cell-cycle dependent manner. It can mediate both cell proliferation and p53-dependent/independent apoptosis. [provided by RefSeq
Specifications
| E2F1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of E2F1. | |
| In PBS (0.09% sodium azide) | |
| NP_005216;Q01094 | |
| E2F1 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S337 of human E2F1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| E2F1 | |
| E2F-1/RBAP1/RBBP3/RBP3 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 1869 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only