Learn More
DUS2L Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant DUS2L.
Brand: Abnova PAB28129.100uL
Description
Dihydrouridine synthase catalyzes reduction of the 5,6-double bond of a uridine residue on the displacement loop of tRNA. The resultant modified base, 5,6-dihydrouridine, appears to increase the conformational flexibility and dynamic motion of tRNA (Kato et al., 2005 [PubMed 15994936]).[supplied by OMIM
Sequence: RLVENDVAGIDVNMGCPKQYSTKGGMGAALLSDPDKIEKILSTLVKGTRRPVTCKIRILPSLEDTLSLVKRIERTGIAAIAVHGRKREERPQHPVSCEVIKAIADTLSIPSpecifications
| DUS2L | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant DUS2L. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| Q9NX74 | |
| DUS2L | |
| Recombinant protein corresponding to amino acids of recombinant DUS2L. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| DUS2L | |
| DUS2/FLJ20399/SMM1/URLC8 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 54920 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only