Learn More
DSCR3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DSCR3.
Brand: Abnova PAB4392.400uL
Description
The region of chromosome 21 between genes CBR and ERG (CBR-ERG region), which spans 2.5Mb on 21q22.2, has been defined by analysis of patients with partial trisomy 21. It contributes significantly to the pathogenesis of many characteristics of Down syndrome, including morphological features, hypotonia, and mental retardation. The DSCR3 (Down syndrome critical region gene 3) gene is found in this region and is predictated to contain eight exons. DSCR3 is expressed in most tissues examined. [provided by RefSeq
Specifications
| DSCR3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of DSCR3. | |
| In PBS (0.09% sodium azide) | |
| NP_006043;O14972 | |
| DSCR3 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human DSCR3. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:50-100) The optimal working dilution should be determined by the end user. | |
| DSCR3 | |
| DCRA/DSCRA/MGC117385 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 10311 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only