missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRAXIN Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DRAXIN.
Brand: Abnova PAB25728.100ug
This item is not returnable.
View return policy
Specifications
| DRAXIN | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| DRAXIN | |
| AGPA3119/PSEC0258/C1orf187/UNQ3119/neucrin | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 374946 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of DRAXIN. | |
| In PBS (0.02% sodium azide) | |
| BAG80561 | |
| DRAXIN | |
| A synthetic peptide corresponding to 17 amino acids at C-terminus of human DRAXIN. | |
| 100 μg | |
| Primary | |
| At least three isoforms of Neucrin are known to exist; this antibody will detect the two shorter isoforms. Neucrin antibody is predicted to not cross-react with RILP. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction