Learn More
DPM3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant DPM3.
Brand: Abnova PAB20750.100uL
Description
Dolichol-phosphate mannose (Dol-P-Man) serves as a donor of mannosyl residues on the lumenal side of the endoplasmic reticulum (ER). Lack of Dol-P-Man results in defective surface expression of GPI-anchored proteins. Dol-P-Man is synthesized from GDP-mannose and dolichol-phosphate on the cytosolic side of the ER by the enzyme dolichyl-phosphate mannosyltransferase. The protein encoded by this gene is a subunit of dolichyl-phosphate mannosyltransferase and acts as a stabilizer subunit of the dolichyl-phosphate mannosyltransferase complex. [provided by RefSeq
Sequence: FHDCEDAARELQSQIQEARADLARRSpecifications
| DPM3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant DPM3. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9P2X0 | |
| DPM3 | |
| Recombinant protein corresponding to amino acids of human DPM3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| DPM3 | |
| MGC125904/MGC125905/MGC34275 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 54344 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only