Learn More
DPEP1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DPEP1.
Brand: Abnova PAB26064.50ug
Description
DPEP1 (EC 3.4.13.11) is a kidney membrane enzyme that hydrolyzes a variety of dipeptides and is implicated in renal metabolism of glutathione and its conjugates, e.g., leukotriene D4 (Kozak and Tate, 1982 [PubMed 6122685]). DPEP1 is responsible for hydrolysis of the beta-lactam ring of antibiotics, such as penem and carbapenem (Campbell et al., 1984 [PubMed 6334084]). Earlier, beta-lactamase enzymes were thought to occur only in bacteria, where their probable function was in protecting the organisms against the action of beta-lactam antibiotics. These antibiotics exhibit selective toxicity against bacteria but virtual inertness against many eukaryotic cells (Adachi et al., 1990 [PubMed 2303490]).[supplied by OMIM
Specifications
| DPEP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of DPEP1. | |
| In PBS (0.09% sodium azide) | |
| P16444 | |
| DPEP1 | |
| A synthetic peptide corresponding to 18 amino acids at N-terminus of human DPEP1. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| DPEP1 | |
| MBD1/MDP/RDP | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 1800 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only