Learn More
DNAJC25-GNG10, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant DNAJC25-GNG10.
Brand: Abnova PAB27473.100uL
Description
This gene represents naturally-occurring mRNAs that are co-transcribed products of the neighboring DNAJC25 and GNG10 genes. These transcripts include the first exon of DNAJC25 and the last two exons of GNG10, resulting in a protein that combines the N-terminus of DNAJC25 and the C-terminus of GNG10. [provided by RefSeq
Sequence: NIKGKEYGEEERLYIIRKSMKMSKSQFDSLEDHQKETFLKRELWIKENYEVYKQEQEEELKKKLANDPRWKRYRRWMKNEGPGRLTFVDDSpecifications
| DNAJC25-GNG10 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant DNAJC25-GNG10. | |
| In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) | |
| DNAJC25-GNG10 | |
| Recombinant protein corresponding to amino acids of human DNAJC25-GNG10. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| DNAJC25-GNG10 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 552891 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only