Learn More
DNAH8, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant DNAH8.
Brand: Abnova PAB22154.100uL
Description
Dyneins are microtubule-associated motor protein complexes composed of several heavy, light, and intermediate chains. Dynein heavy chains (DHCs) are responsible for force production and ATPase activity and contain a highly conserved catalytic domain with 4 P-loop consensus motifs involved in nucleotide binding. Two major classes of dyneins, axonemal and cytoplasmic, have been identified. Axonemal dyneins, found in cilia and flagella, are components of the outer and inner dynein arms attached to the peripheral microtubule doublets. DNAH8 is an outer arm axonemal DHC (Chapelin et al., 1997 [PubMed 9256245]; Neesen et al., 1997 [PubMed 9373155]).[supplied by OMIM
Sequence: ILNHKSKHVEEAVRELISIFEQIYEVKYTGKVGKQSEQRKHVVFGSETEEGENNDYEANIVNEFDTHDKEDEFKKECKEVFAFFSHQLLDSLQKATRLSLDSpecifications
| DNAH8 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant DNAH8. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q96JB1 | |
| DNAH8 | |
| Recombinant protein corresponding to amino acids of human DNAH8. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| DNAH8 | |
| ATPase/FLJ25850/FLJ36115/FLJ36334/hdhc9 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1769 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only