Learn More
DNAAF2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant DNAAF2.
Brand: Abnova PAB20263.100uL
Description
The KTU gene encodes a protein conserved in animals and ciliated unicellular organisms involved in preassembly of dynein arm complexes in species from algae to humans (Omran et al., 2008 [PubMed 19052621]).[supplied by OMIM
Sequence: GEPLCPPLLCNQDKETLTLLIQVPRIQPQSLQGDLNPLWYKLRFSAQDLVYSFFLQFAPENKLSTTEPVISISSNNAVIELAKSPESHGHWREWYYGVNNDSLESpecifications
| DNAAF2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant DNAAF2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9NVR5 | |
| DNAAF2 | |
| Recombinant protein corresponding to amino acids of human DNAAF2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| DNAAF2 | |
| C14orf104/CILD10/KTU/PF13 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 55172 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only