Learn More
DMGDH Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant DMGDH.
Brand: Abnova PAB23001.100uL
Description
This gene encodes an enzyme involved in the catabolism of choline, catalyzing the oxidative demethylation of dimethylglycine to form sarcosine. The enzyme is found as a monomer in the mitochondrial matrix, and uses flavin adenine dinucleotide and folate as cofactors. Mutation in this gene causes dimethylglycine dehydrogenase deficiency, characterized by a fishlike body odor, chronic muscle fatigue, and elevated levels of the muscle form of creatine kinase in serum. [provided by RefSeq
Sequence: TLATDDVDPEGNESIWYNGKVVGNTTSGSYSYSIQKSLAFAYVPVQLSEVGQQVEVELLGKNYPAVIIQEPLVLTEPTRNRLQKSpecifications
| DMGDH | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant DMGDH. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9UI17 | |
| DMGDH | |
| Recombinant protein corresponding to amino acids of human DMGDH. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| DMGDH | |
| DMGDHD/ME2GLYDH | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 29958 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only