missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLK1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DLK1.
Brand: Abnova PAB25633.100ug
This item is not returnable.
View return policy
Specifications
| DLK1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| DLK1 | |
| DLK/FA1/PREF1/Pref-1/ZOG/pG2 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 8788 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of DLK1. | |
| In PBS (0.05% sodium azide) | |
| NP_003827 | |
| DLK1 | |
| A synthetic peptide corresponding to 18 amino acids at C-terminus of human DLK1. | |
| 100 μg | |
| Primary | |
| At least four isoforms of DLK1 are known to exist. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction