missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLGAP5 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DLGAP5.
Brand: Abnova PAB17694.100ug
This item is not returnable.
View return policy
Specifications
| DLGAP5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of DLGAP5. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| Q15398 | |
| DLGAP5 | |
| A synthetic peptide corresponding to internal of human DLGAP5. | |
| RUO | |
| 9787 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
| DLGAP5 | |
| DLG1/DLG7/HURP/KIAA0008 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total DLGAP5 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction