Learn More
DLG2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant DLG2.
Brand: Abnova PAB21384.100uL
Description
This gene encodes a member of the membrane-associated guanylate kinase (MAGUK) family. The encoded protein forms a heterodimer with a related family member that may interact at postsynaptic sites to form a multimeric scaffold for the clustering of receptors, ion channels, and associated signaling proteins. Multiple transcript variants encoding different isoforms have been found for this gene. Additional transcript variants have been described, but their full-length nature is not known. [provided by RefSeq
Sequence: KVGKPTTIYMTDPYGPPDITHSYSPPMENHLLSGNNGTLEYKTSLPPISPGRYSPIPKHMLVDDDYTRPPEPVYSTVNKLCDKPSpecifications
| DLG2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant DLG2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q15700 | |
| DLG2 | |
| Recombinant protein corresponding to amino acids of human DLG2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| DLG2 | |
| DKFZp781D1854/DKFZp781E0954/FLJ37266/MGC131811/PSD-93/PSD93/chapsyn-110 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1740 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only