Learn More
DLD Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DLD.
Brand: Abnova PAB19540.100ug
Description
This gene encodes the L protein of the mitochondrial glycine cleavage system. The L protein, also named dihydrolipoamide dehydrogenase, is also a component of the pyruvate dehydrogenase complex, the alpha-ketoglutarate dehydrogenase complex, and the branched-chain alpha-keto acide dehydrogenase complex. Mutations in this gene have been identified in patients with E3-deficient maple syrup urine disease and lipoamide dehydrogenase deficiency. [provided by RefSeq
Specifications
| DLD | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of DLD. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| DLDH/E3/GCSL/LAD/PHE3 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 1738 | |
| Human, Mouse, Rat | |
| Lyophilized |
| Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (1 ug/mL) Immunocytochemistry (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| DLD | |
| DLD | |
| A synthetic peptide corresponding to amino acids at C-terminus of human DLD. | |
| 100 μg | |
| Primary | |
| The synthetic peptide different from the related mouse and rat sequence by one amino acids. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water and concentration will be 500μ/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only