Learn More
DKKL1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DKKL1.
Brand: Abnova PAB3576.400uL
Description
The dickkopf protein family interacts with the Wnt signaling pathway and its members are characterized by two conserved cysteine-rich domains. This gene encodes a secreted protein that has high similarity to the N-terminus of the dickkopf-3 protein and moderate similarity to that protein's C-terminus though the C-terminal cysteine residues are not conserved. [provided by RefSeq]
Specifications
| DKKL1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of DKKL1. | |
| In PBS (0.09% sodium azide) | |
| NP_055234 | |
| DKKL1 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human DKKL1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| DKKL1 | |
| SGY/SGY-1/SGY1 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 27120 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only