Learn More
DISP1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DISP1.
Brand: Abnova PAB25755.100ug
Description
The pattern of cellular proliferation and differentiation that leads to normal development of embryonic structures often depends upon the localized production of secreted protein signals. Cells surrounding the source of a particular signal respond in a graded manner according to the effective concentration of the signal, and this response produces the pattern of cell types constituting the mature structure. A novel segment-polarity gene known as dispatched has been identified in Drosophila and its protein product is required for normal Hedgehog (Hh) signaling. This gene is one of two human homologs of Drosophila dispatched and, based on sequence identity to its mouse counterpart, the encoded protein may play an essential role in Hh patterning activities in the early embryo. [provided by RefSeq
Specifications
| DISP1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| DISP1 | |
| DISPA/DKFZp434I0428/FLJ43740/MGC104180/MGC13130/MGC16796 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 84976 | |
| Human, Mouse | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of DISP1. | |
| In PBS (0.02% sodium azide) | |
| NP_116279 | |
| DISP1 | |
| A synthetic peptide corresponding to 16 amino acids at N-terminus of human DISP1. | |
| 100 μg | |
| Primary | |
| At least two isoforms of DISP1 are known to exist; this antibody will detect both isoforms. DISP1 antibody is predicted to not cross-react with EMX1. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only