missing translation for 'onlineSavingsMsg'
Learn More

DHX40, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant DHX40.

Brand:  Abnova PAB24320.100uL

Product Code. 16004707

  • £535.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DDX40 is a member of the DExH/D box family of ATP-dependent RNA helicases. RNA helicases catalyze the unwinding of double-stranded RNA and play a role in RNA metabolism, including pre-mRNA splicing, ribosome biogenesis, and organellar gene expression.[supplied by OMIM

Sequence: SVGRTFCTMDGRGSPVHIHPSSALHEQETKLEWIIFHEVLVTTKVYARIVCPIRYEWVRDLLPKLHEFNAHDLSSVARREVREDARRRWTNKENVKQLKDGISKDVLKKMQRRNDDKSISDARARF
Specifications

Specifications

DHX40
Polyclonal
Rabbit polyclonal antibody raised against recombinant DHX40.
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Q8IX18
DHX40
Recombinant protein corresponding to amino acids of human DHX40.
100 μL
Primary
Human
Liquid
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user.
DHX40
ARG147/DDX40/FLJ22060/FLJ34015/PAD
Rabbit
Antigen affinity purification
RUO
79665
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only