Learn More
DGKD Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DGKD.
Brand: Abnova PAB24888.100uL
Description
This gene encodes a cytoplasmic enzyme that phosphorylates diacylglycerol to produce phosphatidic acid. Diacylglycerol and phosphatidic acid are two lipids that act as second messengers in signaling cascades. Their cellular concentrations are regulated by the encoded protein, and so it is thought to play an important role in cellular signal transduction. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq
Specifications
| DGKD | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| DGKD | |
| DGKD | |
| A synthetic peptide corresponding to DGKD. | |
| 100 μL | |
| Primary | |
| DGKD polyclonal antibody detects endogenous levels of DGKD protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of DGKD. | |
| In 1X PBS, pH 7.2 (0.05% sodium azide) | |
| DGKdelta/FLJ26930/KIAA0145/dgkd-2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8527 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only