Learn More
DGCR2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant DGCR2.
Brand: Abnova PAB21078.100uL
Description
Deletions of the 22q11.2 have been associated with a wide range of developmental defects (notably DiGeorge syndrome, velocardiofacial syndrome, conotruncal anomaly face syndrome and isolated conotruncal cardiac defects) classified under the acronym CATCH 22. The DGCR2 gene encodes a novel putative adhesion receptor protein, which could play a role in neural crest cells migration, a process which has been proposed to be altered in DiGeorge syndrome. [provided by RefSeq
Sequence: RFSRKCPTGWHHYEGTASCYRVYLSGENYWDAAQTCQRLNGSLATFSTDQELRFVLAQEWDQPERSFGWKDQRKLWVGYQYVITGRNRSLEGRWEVAFKGSSEVFLPPDPIFASAMSENDNVFCAQLQCFHFPTLRHHDLHSWHAESCYEKSSFLCKRSQTCVDIKDNVVDEGFYFTPKSpecifications
| DGCR2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant DGCR2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P98153 | |
| DGCR2 | |
| Recombinant protein corresponding to amino acids of human DGCR2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| DGCR2 | |
| DGS-C/DKFZp686I1730/IDD/KIAA0163/LAN/SEZ-12 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9993 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only