Learn More
DGCR14 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant DGCR14.
Brand: Abnova PAB28398.100uL
Description
This gene is located within the minimal DGS critical region (MDGCR) thought to contain the gene(s) responsible for a group of developmental disorders. These disorders include DiGeorge syndrome, velocardiofacial syndrome, conotruncal anomaly face syndrome, and some familial or sporadic conotruncal cardiac defects which have been associated with microdeletion of 22q11.2. The encoded protein may be a component of C complex spliceosomes, and the orthologous protein in the mouse localizes to the nucleus. [provided by RefSeq
Sequence: LRVEGSETPYVDRTPGPAFKILEPGRRERLGLKMANEAAAKNRAKKQEALRRVTENLASLTPKGLSPAMSPALQRLVSRTASKYTDRALRASYTPSPARSTHLKTPASGLQTPTSTPAPGSATRTPLTQDPASITDNLSpecifications
| DGCR14 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant DGCR14. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| Q96DF8 | |
| DGCR14 | |
| Recombinant protein corresponding to human DGCR14. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:250-1:500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| DGCR14 | |
| DGCR13/DGS-H/DGS-I/DGSH/DGSI/ES2/Es2el | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8220 | |
| Store at 4 °C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only