Learn More
DES (phospho T76/T77), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of DES.
Brand: Abnova PAB12619.100ug
Description
This gene encodes a muscle-specific class III intermediate filament. Homopolymers of this protein form a stable intracytoplasmic filamentous network connecting myofibrils to each other and to the plasma membrane. Mutations in this gene are associated with desmin-related myopathy, a familial cardiac and skeletal myopathy (CSM), and with distal myopathies. [provided by RefSeq
Specifications
| DES | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of DES. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| CMD1I/CSM1/CSM2/FLJ12025/FLJ39719/FLJ41013/FLJ41793 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody recognizes ∼55 KDa of human DES at dual phosphorylated threonine (Thr76/Thr77). It does not recognize non-phosphorylated peptides confirmed by dot blot. | |
| Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| DES | |
| DES | |
| Synthetic phosphopeptide corresponding to residues surrounding T76/T77 of human DES. | |
| RUO | |
| 1674 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only