Learn More
DEGS1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DEGS1.
Brand: Abnova PAB9854.100ug
Description
This gene encodes a member of the membrane fatty acid desaturase family which is responsible for inserting double bonds into specific positions in fatty acids. This protein contains three His-containing consensus motifs that are characteristic of a group of membrane fatty acid desaturases. It is predicted to be a multiple membrane-spanning protein localized to the endoplasmic reticulum. Overexpression of this gene inhibited biosynthesis of the EGF receptor, suggesting a possible role of a fatty acid desaturase in regulating biosynthetic processing of the EGF receptor. Two splice variants have been identified. [provided by RefSeq
Specifications
| DEGS1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of DEGS1. | |
| In PBS (0.08% sodium azide) | |
| DEGS/DES1/Des-1/FADS7/MGC5079/MIG15/MLD | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| DEGS1 | |
| DEGS1 | |
| A synthetic peptide corresponding to human DEGS1. | |
| RUO | |
| 8560 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only