Learn More
DDX58 (phospho S8), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of DDX58.
Brand: Abnova PAB15905.100ug
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases which are implicated in a number of cellular processes involving RNA binding and alteration of RNA secondary structure. This gene encodes a protein containing RNA helicase-DEAD box protein motifs and a caspase recruitment domain (CARD). It is involved in viral double-stranded (ds) RNA recognition and the regulation of immune response. [provided by RefSeq
Specifications
| DDX58 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of DDX58. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| DKFZp434J1111/DKFZp686N19181/FLJ13599/RIG-I | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| DDX58 | |
| DDX58 | |
| Synthetic phosphopeptide corresponding to residues surrounding S8 of human DDX58. | |
| RUO | |
| 23586 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only