Learn More
DDX4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DDX4.
Brand:  Abnova PAB2479.400uL
Product Code. 16085007
  • £332.00 / 400µL
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a homolog of VASA proteins in Drosophila and several other species. The gene is specifically expressed in the germ cell lineage in both sexes and functions in germ cell development. [provided by RefSeq
Specifications
Specifications
DDX4
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of DDX4.
In PBS (0.09% sodium azide)
NP_077726;Q9NQI0
DDX4
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human DDX4.
400 μL
Primary
Human
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Western Blot (1:1000) The optimal working dilution should be determined by the end user.
DDX4
MGC111074/VASA
Rabbit
Ammonium sulfate precipitation
RUO
54514
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.