Learn More
DDX3X Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DDX3X.
Brand: Abnova PAB13187.100ug
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which interacts specifically with hepatitis C virus core protein resulting a change in intracellular location. This gene has a homolog located in the nonrecombining region of the Y chromosome. The protein sequence is 91% identical between this gene and the Y-linked homolog. [provided by RefSeq
Specifications
| DDX3X | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of DDX3X. | |
| In PBS (0.02% sodium azide) | |
| AAC34298 | |
| DDX3X | |
| A synthetic peptide corresponding to N-terminus 16 amino acids of human DDX3X. | |
| RUO | |
| 1654 | |
| Human, Mouse | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| DDX3X | |
| DBX/DDX14/DDX3/HLP2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| DDX3 antibody will detect both DDX3 and DBY. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only