Learn More
DDX25, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant DDX25.
Brand: Abnova PAB21279.100uL
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of the DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a member of this family. The encoded protein is a gonadotropin-regulated and developmentally expressed testicular RNA helicase. It may serve to maintain testicular functions related to steroidogenesis and spermatogenesis. [provided by RefSeq
Sequence: NSHFSNLSQPRKNLWGIKSTAVRNIDGSINNINEDDEEDVVDLAANSLLNKLIHQSLVESSHRVEVLQKDPSSPSpecifications
| DDX25 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant DDX25. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9UHL0 | |
| DDX25 | |
| Recombinant protein corresponding to amino acids of human DDX25. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| DDX25 | |
| GRTH | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 29118 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only