missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DAP.
Brand: Abnova PAB27443.100ug
This item is not returnable.
View return policy
Description
This gene encodes a basic, proline-rich, 15kDa protein. The protein acts as a positive mediator of programmed cell death that is induced by interferon-gamma. [provided by RefSeq
Specifications
| DAP | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (5 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| DAP | |
| MGC99796 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 1611 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of DAP. | |
| In PBS (0.02% sodium azide) | |
| NP_004385 | |
| DAP | |
| A synthetic peptide corresponding to 19 amino acids at C-terminus of human DAP. | |
| 100 μg | |
| Primary | |
| DAP antibody is human, mouse and rat reactive. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction