Learn More
DAD1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DAD1.
Brand: Abnova PAB12919.100ug
Description
DAD1, the defender against apoptotic cell death, was initially identified as a negative regulator of programmed cell death in the temperature sensitive tsBN7 cell line. The DAD1 protein disappeared in temperature-sensitive cells following a shift to the nonpermissive temperature, suggesting that loss of the DAD1 protein triggered apoptosis. DAD1 is believed to be a tightly associated subunit of oligosaccharyltransferase both in the intact membrane and in the purified enzyme, thus reflecting the essential nature of N-linked glycosylation in eukaryotes. [provided by RefSeq
Specifications
| DAD1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of DAD1. | |
| In PBS (0.02% sodium azide) | |
| AAH09798 | |
| DAD1 | |
| A synthetic peptide corresponding to C-terminus 14 amino acids of human DAD1. | |
| RUO | |
| 1603 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| DAD1 | |
| OST2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only