Learn More
DACH2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant DACH2.
Brand: Abnova PAB19960.100uL
Description
This gene is one of two genes which encode a protein similar to the Drosophila protein dachshund, a transcription factor involved in cell fate determination in the eye, limb and genital disc of the fly. The encoded protein contains two characteristic dachshund domains: an N-terminal domain responsible for DNA binding and a C-terminal domain responsible for protein-protein interactions. This gene is located on the X chromosome and is subject to inactivation by DNA methylation. The encoded protein may be involved in regulation of organogenesis and myogenesis, and may play a role in premature ovarian failure. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: ITAAAMAEAMKLQKMKLMAMNTLQGNGSQNGTESEPDDLNSNTGGSESSWDKDKMQSPFAAPGPQHGIAHAALAGQPGIGGAPTLNPLQQNHLLTNRLDLPFMMMPHPLLPVSLPPASVSpecifications
| DACH2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant DACH2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q96NX9 | |
| DACH2 | |
| Recombinant protein corresponding to amino acids 215-333 of human DACH2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| DACH2 | |
| FLJ31391/MGC138545 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 117154 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only