Learn More
CYSLTR1 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Description
The cysteinyl leukotrienes LTC4, LTD4, and LTE4 are important mediators of human bronchial asthma. Pharmacologic studies have determined that cysteinyl leukotrienes activate at least 2 receptors, the protein encoded by this gene and CYSLTR2. This encoded receptor is a member of the superfamily of G protein-coupled receptors. Activation of this receptor by LTD4 results in contraction and proliferation of smooth muscle, oedema, eosinophil migration and damage to the mucus layer in the lung. [provided by RefSeq
Specifications
Specifications
| Antigen | CYSLTR1 |
| Applications | Immunohistochemistry (PFA fixed) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic peptide of CYSLTR1. |
| Dilution | Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (6-14 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS (0.09% sodium azide) |
| Gene | CYSLTR1 |
| Gene Accession No. | Q9Y271 |
| Gene Alias | CYSLT1/CYSLT1R/CYSLTR/HG55/HMTMF81/MGC46139 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.