Learn More
CYP39A1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CYP39A1.
Brand: Abnova PAB17530.100ug
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein is involved in the conversion of cholesterol to bile acids. Its substrates include the oxysterols 25-hydroxycholesterol, 27-hydroxycholesterol and 24-hydroxycholesterol. [provided by RefSeq
Specifications
| CYP39A1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CYP39A1. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| Q9NYL5 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total CYP39A1 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunofluorescence (1:500-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
| CYP39A1 | |
| CYP39A1 | |
| A synthetic peptide corresponding to internal of human CYP39A1. | |
| RUO | |
| 51302 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only