Learn More
CXCR7 Rabbit anti-Bovine, Dog, Hamster, Horse, Human, Mouse, Pig, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CXCR7.
Marke: Abnova PAB26187.50ug
Beschreibung
This gene encodes a member of the G-protein coupled receptor family. Although this protein was earlier thought to be a receptor for vasoactive intestinal peptide (VIP), it is now considered to be an orphan receptor, in that its endogenous ligand has not been identified. The protein is also a coreceptor for human immunodeficiency viruses (HIV). Translocations involving this gene and HMGA2 on chromosome 12 have been observed in lipomas. [provided by RefSeq
Spezifikation
| CXCR7 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CXCR7. | |
| In PBS (0.09% sodium azide) | |
| P25106 | |
| CXCR7 | |
| A synthetic peptide corresponding to 19 amino acid at extracellular domain of human CXCR7. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Flow Cytometry, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| CXCR7 | |
| CMKOR1/GPR159/RDC1 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 57007 | |
| Bovine, Canine, Equine, Hamster, Human, Mouse, Pig, Rabbit, Rat | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only