Learn More
CXCL9, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CXCL9.
Brand: Abnova PAB19516.100ug
Description
The function of this gene has not been specifically defined; however, it is thought to be involved in T cell trafficking. This gene has been localized to 4q21 with INP10, which is also a member of the chemokine family of cytokines. [provided by RefSeq
Specifications
| CXCL9 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CXCL9. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| CMK/Humig/MIG/SCYB9/crg-10 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 4283 | |
| Human | |
| Lyophilized |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CXCL9 | |
| CXCL9 | |
| A synthetic peptide corresponding to amino acids at N-terminus of human CXCL9. | |
| 100 μg | |
| Primary | |
| It identical to the rat and mouse sequences. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water and concentration will be 500μ/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only