Learn More
Cx3cl1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Cx3cl1.
Brand: Abnova PAB19482.100ug
This item is not returnable.
View return policy
Specifications
| Cx3cl1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Cx3cl1. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| AB030188/ABCD-3/AI848747/CX3C/Cxc3/D8Bwg0439e/Scyd1 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 20312 | |
| Mouse, Rat | |
| Lyophilized |
| Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Cx3cl1 | |
| Cx3cl1 | |
| A synthetic peptide corresponding to amino acids at N-terminus of mouse Cx3cl1. | |
| 100 μg | |
| Primary | |
| It identical to the related human and rat sequence. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water and concentration will be 500μ/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |