missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUL2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CUL2.
Brand: Abnova PAB10036.100uL
This item is not returnable.
View return policy
Specifications
| CUL2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CUL2. | |
| In antiserum (0.01% sodium azide) | |
| NP_003582 | |
| CUL2 | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 733-745 of human CUL2. | |
| RUO | |
| 8453 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:10000) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| CUL2 | |
| MGC131970 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction